missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAVS Antibody, R&D Systems™

Mouse Monoclonal Antibody
209.00 € - 413.00 €
Specifications
| Antigen | MAVS |
|---|---|
| Clone | 981240 |
| Dilution | Immunohistochemistry 1-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715968
![]() |
R&D Systems
MAB9694-SP |
25 μg |
209.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18755179
![]() |
R&D Systems
MAB9694-100 |
100 μg |
413.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAVS Monoclonal specifically detects MAVS in Human samples. It is validated for Immunohistochemistry.Specifications
| MAVS | |
| Immunohistochemistry 1-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| Q7Z434 | |
| 57506 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 981240 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CARD adapter inducing interferon beta, CARD adapter inducing interferon-beta, CARD adaptor inducing IFN-beta, CARDIF, DKFZp547C224, DKFZp666M015, FLJ27482, IFN-B promoter stimulator 1, Interferon beta promoter stimulator protein 1, interferon-beta promoter stimulator protein 1, IPS-1, IPS-1FLJ41962, IPS1MGC3260, KIAA1271FLJ35386, mitochondrial antiviral signaling protein, mitochondrial antiviral-signaling protein, mitochondrial viral signaling protein, Putative NF-kappa-B-activating protein 031N, virus-induced signaling adapter variant 1b, virus-induced signaling adaptor variant 1a, Virus-induced-signaling adapter, VISA, VISAFLJ38051 | |
| E.coli-derived recombinant human MAVS Arg37-Pro513 Accession # Q7Z434 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MAVS in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title