missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCAM/CD146 Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
234.00 € - 421.00 €
Specifications
| Antigen | MCAM/CD146 |
|---|---|
| Clone | 128018 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723947
![]() |
R&D Systems
FAB932F-025 |
25 Tests |
234.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18794477
![]() |
R&D Systems
FAB932F-100 |
100 Tests |
421.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCAM/CD146 Monoclonal specifically detects MCAM/CD146 in Human samples. It is validated for Flow Cytometry.Specifications
| MCAM/CD146 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD146, CD146 antigen, cell surface glycoprotein MUC18, Cell surface glycoprotein P1H12, L-Gicerin, melanoma adhesion molecule, melanoma cell adhesion moleculeS-endo 1 endothelial-associated antigen, Melanoma-associated antigen A32, Melanoma-associated antigen MUC18, MUC18, MUC18Gicerin | |
| MCAM | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human MCAM/CD146 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhNCAM-L1, recombinant mouse (rm)OCAM, or rmMAdCAM-1. |
| 128018 | |
| Flow Cytometry | |
| Fluorescein | |
| Mouse | |
| Human | |
| AAA20824 | |
| 4162 | |
| Mouse myeloma cell line NS0-derived recombinant human MCAM/CD146 Val24-Gly559 Accession # AAA20824 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title