missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human mCherry Antibody, R&D Systems™

Rabbit Monoclonal Antibody
208.00 € - 534.00 €
Specifications
| Antigen | mCherry |
|---|---|
| Clone | 2783C |
| Dilution | Western Blot 0.25 ug/mL, Simple Western 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL |
| Applications | Western Blot, Flow Cytometry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18791467
![]() |
R&D Systems
MAB11041-SP |
25 μg |
208.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18781527
![]() |
R&D Systems
MAB11041-100 |
100 μg |
534.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
mCherry Monoclonal antibody specifically detects mCherry in Human samples. It is validated for Western Blot, Western Blot, Flow Cytometry, ImmunocytochemistrySpecifications
| mCherry | |
| Western Blot 0.25 ug/mL, Simple Western 25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| Human mCherry synthetic peptide | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 2783C | |
| Western Blot, Flow Cytometry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| DSRED, red fluorescent protein mCherry, Red Fluoroscent Protein | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title