missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCHR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
176.00 € - 453.00 €
Specifications
| Antigen | MCHR1 |
|---|---|
| Clone | 1032150 |
| Dilution | Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794617
![]() |
R&D Systems
MAB79382-SP |
25 μg |
176.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18770987
![]() |
R&D Systems
MAB79382-100 |
100 μg |
453.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCHR1 Monoclonal antibody specifically detects MCHR1 in Human samples. It is validated for Immunohistochemistry, ImmunocytochemistrySpecifications
| MCHR1 | |
| Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| G protein-coupled receptor 24, GPR24, GPR24SLC-1, G-protein coupled receptor 24, MCH receptor 1, MCH1R, MCH-1R, MCH1RMCHR-1, MCH-R1, melanin-concentrating hormone receptor 1, MGC32129, SLC1, SLC1MCHR, Somatostatin receptor-like protein | |
| E. coli-derived human MCHR1, Met1-Pro113, Accession # Q99705 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1032150 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q99705 | |
| 2847 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title