missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mcl-1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8281-SP
This item is not returnable.
View return policy
Description
Mcl-1 Polyclonal specifically detects Mcl-1 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| Mcl-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| bcl2-L-3, BCL2L3, BCL2L3MGC104264, Bcl-2-like protein 3, Bcl-2-related protein EAT/mcl1, EAT, induced myeloid leukemia cell differentiation protein Mcl-1, Mcl1, Mcl-1, mcl1/EAT, MCL1-ES, MCL1L, MCL1S, MGC1839, myeloid cell leukemia ES, myeloid cell leukemia sequence 1 (BCL2-related), TM | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4170 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoassay | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 20 ug/mL, Knockout Validated | |
| Q07820 | |
| MCL1 | |
| E. coli-derived recombinant human Mcl-1 Val147-Gly219 Accession # Q07820 | |
| 25 μg | |
| Primary | |
| Detects human Mcl-1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction