missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MICA Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB1300G-025
This item is not returnable.
View return policy
Description
MICA Monoclonal specifically detects MICA in Human samples. It is validated for Flow Cytometry.
Specifications
| MICA | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| MICA | |
| BaF3 mouse pro-B cell line transfected with human MICA | |
| 25 Tests | |
| Primary | |
| Detects human MICA in direct ELISAs and Western blots. In direct ELISAs, this antibody does not cross-react with recombinant human MICB. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 159227 | |
| Flow Cytometry 5 uL/10^6 cells | |
| FLJ60820, MGC111087, PERB11.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 100507436 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction