Learn More
Human Mind Bomb 2/MIB2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB72891
Product Code. 18129640
  • 412.00 € / 100µg
Description
Mind Bomb 2/MIB2 Monoclonal specifically detects Mind Bomb 2/MIB2 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mind Bomb 2/MIB2
Monoclonal
LYOPH
Western Blot 2 ug/mL
Q96AX9
MIB2
E. coli-derived recombinant human Mind Bomb 2/MIB2 Ala418-Lys520 Accession # Q96AX9
100 μg
Primary
Detects human Mind Bomb 2/MIB2 in direct ELISAs and Western blots.
Human
Purified
Western Blot
739505
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
E3 ubiquitin-protein ligase MIB2, EC 6.3.2, EC 6.3.2.-, EC 6.3.2.13, FLJ20648, FLJ41406, MIB2, Mind bomb homolog 2, Mind Bomb-2, mindbomb homolog 2 (Drosophila), Novel zinc finger protein, novelzin, Putative NF-kappa-B-activating protein 002N, SKD, Skeletrophin, Zinc finger ZZ type with ankyrin repeat domain protein 1, ZZ type with ankyrin repeat domain 1, ZZANK1, ZZZ5
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
142678
Sterile PBS to a final concentration of 0.5 mg/mL.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b