missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-1 Antibody, R&D Systems™

Goat Monoclonal Antibody
Brand: R&D Systems MAB9011-100
This item is not returnable.
View return policy
Beschreibung
MMP-1 Monoclonal specifically detects MMP-1 in Human samples. It is validated for Western Blot, Knockout Validated.
Spezifikation
| MMP-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP1, MMP-1 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-1 Phe20-Asn469 Accession # P03956 | |
| 100 μg | |
| Primary | |
| Detects human MMP-1 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, KnockDown | |
| 40013F | |
| Western Blot 5 ug/mL, Knockout Validated | |
| P03956 | |
| Goat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 4312 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur