missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BMPR-IB/ALK-6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB505-500
This item is not returnable.
View return policy
Description
BMPR-IB/ALK-6 Monoclonal specifically detects BMPR-IB/ALK-6 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BMPR-IB/ALK-6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMPR1B | |
| Mouse myeloma cell line NS0-derived recombinant human BMPR-IB/ALK-6 | |
| 500 μg | |
| Primary | |
| Detects human and mouse BMPR-IB/ALK-6 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) BMPR-IB and no cross-reactivity with recombinant human (rh) BMPR-IA, rmBMPR-IA, or rhBMPR-II is observed. | |
| Human, Mouse | |
| Ascites |
| Western Blot | |
| 88614 | |
| Western Blot 1 ug/mL | |
| activin receptor-like kinase 6, ALK-6, ALK6CDw293, BMP type-1B receptor, BMPR1B, BMPR-1B, BMPRIB, bone morphogenetic protein receptor type-1B, bone morphogenetic protein receptor, type IB, CDw293, CDw293 antigen, EC 2.7.11, EC 2.7.11.30, serine/threonine receptor kinase | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 658 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction