missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse CD45R / B220 Rat, Clone: RA3-6B2, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB41-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15951978

  • 94.25 € / 100µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Human/Mouse CD45R / B220
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Ptprc
Protein A/G Purified
RUO
19264
IgG2a κ
Flow Cytometry
RA3-6B2
Unconjugated
NP_001255215
Rat
100 μg
Primary
Store at 2-4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers