missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD45R / B220 Rat, Qfluor 630, Clone: RA3-6B2, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB41-QF630-100ug
This item is not returnable.
View return policy
Specifications
| Human/Mouse CD45R / B220 | |
| Monoclonal | |
| 0.2mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| Ptprc | |
| Protein A/G Purified | |
| RUO | |
| 19264 | |
| IgG2a κ |
| Flow Cytometry | |
| RA3-6B2 | |
| Qfluor 630 | |
| NP_001255215 | |
| Rat | |
| 100 μg | |
| Primary | |
| Store at 2-4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction