missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse COX4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB6980
This item is not returnable.
View return policy
Description
COX4 Monoclonal specifically detects COX4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| COX4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q96KJ9 | |
| COX4I2 | |
| E.coli-derived recombinant human COX4-I2 Asn39-Lys171 Accession # Q96KJ9 | |
| 100 μg | |
| Primary | |
| Detects human COX4-I2 in direct ELISAs. In Western blots, 100% cross-reactivity with recombinant human (rh) COX4-I1 and no cross-reactivity with rhCOX-1 or rhCOX-2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 673803 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COX IV-1, COX4-1, COX4-2, COX4B, COX4COXIV, COX4I1, COX4I2, COX4L2, COXIV-2, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1327 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction