Learn More
Human/Mouse CXCL12/SDF-1 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 38 publications
Brand:  R&D Systems MAB310-500
 View more versions of this product
Product Code. 18715148
  • 2140.00 € / 500µg
Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CXCL12/SDF-1
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSF, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF1, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
6387
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Neutralization
79014
Neutralization
Q6ICW0
CXCL12
E. coli-derived recombinant human CXCL12/SDF-1 alpha Lys22-Lys89 Accession # Q6ICW0
500 μg
Primary
Detects human and mouse CXCL12/SDF-1 alpha and human CXCL12/SDF-1 beta .
Human, Mouse
Supernatant