missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CXCR7/RDC-1 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB4227P-025
This item is not returnable.
View return policy
Description
CXCR7/RDC-1 Monoclonal specifically detects CXCR7/RDC-1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CXCR7/RDC-1 | |
| Monoclonal | |
| R-PE | |
| P25106 | |
| CXCR7 | |
| Human CXCR7 encoding plasmid Accession # P25106 | |
| 25 Tests | |
| 57007 | |
| Human, Mouse | |
| Purified |
| Flow Cytometry | |
| 11G8 | |
| Flow Cytometry 10 uL/10^6 cells | |
| ACKR3, chemokine (C-X-C motif) receptor 7, Chemokine orphan receptor 1CMKOR1C-X-C chemokine receptor type 7, CMKOR1, CXCR7, CXC-R7, CXCR-7, G protein-coupled receptor, GPR159, GPR159G-protein coupled receptor 159, RDC-1, RDC1G-protein coupled receptor RDC1 homolog | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CXCR7/RDC-1 in flow cytometry of five distinct human CXCR7 transfectants, but not their respective parental lines. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction