Learn More
Human/Mouse FoxP3 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Monoclonal Antibody
Brand:  R&D Systems FPK8214P-100
 View more versions of this product
Product Code. 18783148
  • 549.00 € / 100 tests
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
FoxP3
Monoclonal
R-PE
Q9BZS1
FOXP3
E. coli-derived recombinant human FoxP3 Met1-Leu71 Accession # Q9BZS1
100 Tests
50943
Human, Mouse
IgG
Flow Cytometry
1054C
Flow Cytometry 10 uL/10^6 cells
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked
Rabbit
Affinity Purified
Primary
Detects human and mouse FoxP3 in flow cytometry.
Store the unopened product at 2 to 8 degreesC. Do not use past expiration date. Protect from light.