Learn More
Human/Mouse IL-12 R beta 2 PerCP-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems FAB1959C-100
 View more versions of this product
Product Code. 18788137
  • 552.00 € / 100 tests
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-12 R beta 2
Monoclonal
PerCP
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
3595
Human, Mouse
Purified
Flow Cytometry
305719
Flow Cytometry 10 uL/10^6 cells
Q99665
IL12RB2
Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665
100 Tests
Primary
Detects human and mouse IL-12 R beta 2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-1 R beta 1 is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG1