missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Indian Hedgehog/Ihh N-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18712909
Change view
Click to view available options
Quantity:
25 μg
500 μg
Unit Size:
25µg
500µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18712909 25 μg 25µg
18190211 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18712909 Supplier R&D Systems Supplier No. MAB1705SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Indian Hedgehog/Ihh Monoclonal specifically detects Indian Hedgehog/Ihh in Human, Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Indian Hedgehog/Ihh
Applications Western Blot
Classification Monoclonal
Clone 187515
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P97812
Gene Alias BDA1, BDA1Indian hedgehog homolog (Drosophila), HHG2, HHG-2, Ihh, Indian hedgehog, Indian hedgehog (Drosophila) homolog, Indian hedgehog homolog, indian hedgehog protein
Gene Symbols IHH
Host Species Rat
Immunogen E. coli-derived recombinant mouse Indian Hedgehog Cys28-Gly202 Accession # P97812
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3549
Test Specificity Detects mouse Indian Hedgehog (Ihh) in direct ELISAs and Western blots. This antibody is specific for the N-terminal peptide of mouse Ihh (amino acids (aa) 66-240). In Western blots, this antibody does not cross-react with recombinant mouse (rm) Dhh (aa 23-198 or aa 199-396), rmIhh (aa 241-449) and has less than 25% cross-reactivity with rhShh (aa 24-197), or rmShh (aa 25-198 or aa 199-437).
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.