missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse LMO4 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF3889
This item is not returnable.
View return policy
Description
LMO4 Polyclonal specifically detects LMO4 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| LMO4 | |
| Polyclonal | |
| Unconjugated | |
| P61968 | |
| LMO4 | |
| E. coli-derived recombinant human LMO4 Met1-Cys165 Accession # P61968 | |
| 100 μg | |
| Primary | |
| Detects human and mouse LMO4 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Breast tumor autoantigen, Crp3, Etohi4, LIM domain only 4, LIM domain only protein 4, LIM domain transcription factor LMO4, LMO-4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8543 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction