missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Myeloperoxidase/MPO Antibody, R&D Systems™

Description
Myeloperoxidase/MPO Polyclonal specifically detects Myeloperoxidase/MPO in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
Specifications
| Antigen | Myeloperoxidase/MPO |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL |
| Gene Accession No. | P11247 |
| Gene Alias | EC 1.11.1, EC 1.11.1.7, MPO, myeloperoxidase |
| Gene Symbols | MPO |
| Host Species | Goat |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?