missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat CTGF/CCN2 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB91901-100
This item is not returnable.
View return policy
Description
CTGF/CCN2 Monoclonal specifically detects CTGF/CCN2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| CTGF/CCN2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CCN2, CCN2IGFBP-8, connective tissue growth factor, CTGRP, Fisp12, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP-8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2 | |
| E. coli-derived recombinant human CTGF/CCN2 Glu247-Ala349 Accession # P29279 | |
| 100 μg | |
| Primary | |
| Detects human CTGF/CCN2 in direct ELISAs. Detects human, mouse, and rat CTGF/CCN2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 2191A | |
| Western Blot 2 ug/mL | |
| P29279 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1490 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction