missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Ezrin Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB72391-100
This item is not returnable.
View return policy
Description
Ezrin Monoclonal specifically detects Ezrin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.
Specifications
| Ezrin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
| CVIL, CVL, Cytovillin, cytovillin 2, DKFZp762H157, EZR, ezrin, FLJ26216, p81, VIL2, VIL2MGC1584, Villin 2, villin 2 (ezrin), villin-2 | |
| E. coli-derived recombinant human Ezrin Lys438-Arg562 Accession # P15311 | |
| 100 μg | |
| Primary | |
| Detects human Ezrin in direct ELISAs and detects human, mouse, and rat Ezrin in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
| 2086C | |
| Western Blot 0.2 ug/mL, Immunohistochemistry 1-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 3-25 ug/mL | |
| P15311 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7430 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction