missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FKBP12/FKBP12.6 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4174-SP
This item is not returnable.
View return policy
Description
FKBP12/FKBP12.6 Monoclonal specifically detects FKBP12/FKBP12.6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| FKBP12/FKBP12.6 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry | |
| 426227 | |
| Western Blot 1 μg;/mL, Immunohistochemistry 5-25 μg;/mL | |
| P68106 | |
| E. coli-derived recombinant human FKBP12.6, Gly2-Glu108, Accession No. P68106 | |
| 25 μg | |
| Detects human FKBP12.6 in direct ELISAs. Detects human, mouse, and rat FKBP12 and FKBP12.6 in Western blots. | |
| Human, Mouse, Rat | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction