missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GSK-3 alpha/beta Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2157-SP
This item is not returnable.
View return policy
Description
GSK-3 alpha/beta Polyclonal specifically detects GSK-3 alpha/beta in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| GSK-3 alpha/beta | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK3 alpha, GSK-3 alpha, GSK3 beta, GSK3A, GSK3B | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2931 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P49841 | |
| GSK3A | |
| E. coli-derived recombinant human GSK-3 beta Accession # P49841 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat GSK-3 beta and the closely related GSK-3 alpha . | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction