missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Kynurenine 3-Monooxygenase/KMO Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB8050-100
This item is not returnable.
View return policy
Description
Kynurenine 3-Monooxygenase/KMO Monoclonal specifically detects Kynurenine 3-Monooxygenase/KMO in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Kynurenine 3-Monooxygenase/KMO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. | |
| KMO, Kynurenine 3Monooxygenase, kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Kynurenine 3-Monooxygenase/KMO, Asp2-Leu441, Accession # O15229 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| 2493A | |
| Western Blot 1 ug/mL, Immunohistochemistry 3-25 ug/mL | |
| O15229 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 8564 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction