missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Slit3 Antibody, R&D Systems™

Description
Slit3 Polyclonal specifically detects Slit3 in Human, Mouse, Rat samples. It is validated for Western Blot, Neutralization.
Specifications
Specifications
| Antigen | Slit3 |
| Applications | Immunohistochemistry, Neutralization |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 5 ug/mL, Neutralization |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q9WVB4 |
| Gene Alias | KIAA0814, MEGF5, MEGF5Multiple epidermal growth factor-like domains protein 5, Multiple EGF-like domains protein 5, SLIL2, SLIL2FLJ10764, slit homolog 3 (Drosophila), slit homolog 3 protein, SLIT1, slit2, Slit-3slit (Drosophila) homolog 3 |
| Gene Symbols | SLIT3 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?