missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SOX2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2018
This item is not returnable.
View return policy
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-reported.
Specifications
| SOX2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 4 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-reported | |
| P48431 | |
| SOX2 | |
| E. coli-derived recombinant human SOX2 Gly135-Met317 Accession # P48431 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat SOX2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 245610 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6657 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction