missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SOX2 Antibody, R&D Systems™
Description
SOX2 Monoclonal specifically detects SOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
Specifications
Specifications
| Antigen | SOX2 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | 245610R |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells |
| Formulation | Supplied as a solution in PBS. *Small pack size (SP) is supplied as a 0.2 um filtered solution in PBS. with No Preservative |
| Gene Accession No. | P48431 |
| Gene Alias | ANOP3, MCOPS3, MGC2413, SRY (sex determining region Y)-box 2, SRY-related HMG-box gene 2, transcription factor SOX2, transcription factor SOX-2 |
| Gene Symbols | SOX2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?