missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse/Rat Syntaxin 1A Antibody, R&D Systems™

Product Code. 18072222
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072222 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072222 Supplier R&D Systems Supplier No. AF7237
  • ONLINE EXCLUSIVE PRICE
    538.16 €
    473.55 € / 100µg
    Save 64.61 €
    12% Off
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Goat Polyclonal Antibody

Syntaxin 1A Polyclonal specifically detects Syntaxin 1A in Human∣Mouse∣Rat samples. It is validated for Immunocytochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Syntaxin 1A
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q16623
Gene Alias Brain Syntaxin, HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1A, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A
Gene Symbols STX1A
Host Species Goat
Immunogen E. coli-derived recombinant human Syntaxin 1A Met1-Leu165 Accession # Q16623
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6804
Test Specificity Detects human, mouse, and rat Syntaxin 1A in Western blots and detects recombinant human Syntaxin 1A in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Syntaxin 1B2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Product Type Antibody
Isotype IgG
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.