Learn More
Human/Mouse/Rat Vimentin Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB21052-100
 View more versions of this product
Product Code. 18710788
  • 468.00 € / 100µg
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Knockout Validated, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vimentin
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
FLJ36605, VIM, vimentin
E.coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670
100 μg
7431
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoassay, In Situ Hybridization (ISH)
979517
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated 5-15 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL
P08670
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human Vimentin in direct ELISAs. Detects human, mouse, and rat Vimentin in Western blots.
Human, Mouse, Rat
Supernatant