missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human and Mouse Silencer™ siRNA Library, In Plate

Product Code. 10176614
Change view
Click to view available options
Quantity:
1 each
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10176614 1 each Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10176614 Supplier Thermo Scientific™ Supplier No. AM81830
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs, and when available, Silencer™ Validated siRNAs, in 96-well or 384-well plates.

Silencer™ siRNA Libraries feature combinations of Silencer™ Pre-designed siRNAs, and when available, Silencer™ Validated siRNAs, in 96-well or 384-well plates.

TRUSTED_SUSTAINABILITY

Specifications

Product Line Ambion
Product Type siRNA Library
Quantity 1 each
Format 96- or 384-well Plate
RNAi Type siRNA
Species Human, Mouse
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.