missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse TL1A/TNFSF15 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB7441-SP
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human, Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 1 ug/mL | |
| NP_796345.3 | |
| TNFSF15 | |
| E. coli-derived recombinant mouse TL1A/TNFSF15 Ile94-Leu270 Accession # NP_796345.3 | |
| 25 μg | |
| Primary | |
| Detects mouse TL1A/TNFSF15 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human TL1A. | |
| Human, Mouse | |
| Purified |
| Western Blot, Neutralization | |
| 293327 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, TNFSF15, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI, VEGI192A, VEGITNF ligand-related molecule 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9966 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction