Learn More
Human/Mouse TLR9 Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB36581-SP
 View more versions of this product
Product Code. 18737848
  • 214.00 € / 25µg
Description
TLR9 Monoclonal specifically detects TLR9 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TLR9
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
CD289, CD289 antigen, toll-like receptor 9
Human TLR9 synthetic peptide Accession # Q9NR96
25 μg
54106
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Immunohistochemistry, Immunocytochemistry
980915
Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 3-25 ug/mL
Q9NR96
Mouse
Protein A or G purified from hybridoma culture supernatant
Primary
Detects human TLR9 in direct ELISAs.
Human, Mouse
Supernatant