missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Human Multi-tissue Tissue MicroArray (Normal)

Product Code. 18616930 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18616930 5 Slides Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18616930 Supplier Novus Biologicals™ Supplier No. NBP278070
  • ONLINE EXCLUSIVE PRICE
    391.00 €
    369.60 € / Pack of 5
    Save 21.40 €
    5% Off
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The panel contains 5 to 8 different tissues on one slide.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Slides
Species Human
For Use With (Application) Please bake slides at 60C for 30 minutes before use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.