missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NCK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7008
This item is not returnable.
View return policy
Description
NCK1 Monoclonal specifically detects NCK1 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| NCK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P16333 | |
| NCK1 | |
| E. coli-derived recombinant human NCK1 Met1-Ser377 Accession # P16333 | |
| 100 μg | |
| Primary | |
| Detects human NCK1 in direct ELISAs. | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 714506 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cytoplasmic protein NCK1, MGC12668, NCK adaptor protein 1melanoma NCK protein, nck-1, NCKalpha, NCKNCK tyrosine kinase, non-catalytic region of tyrosine kinase, SH2/SH3 adaptor protein NCK-alpha | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4690 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction