missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NF-H Antibody, R&D Systems™

Mouse Monoclonal Antibody
176.00 € - 470.00 €
Specifications
| Antigen | NF-H |
|---|---|
| Clone | 1016103 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18740457
![]() |
R&D Systems
MAB31082-SP |
25 μg |
176.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18763167
![]() |
R&D Systems
MAB31082-100 |
100 μg |
470.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NF-H Monoclonal antibody specifically detects NF-H in Human samples. It is validated for Western BlotSpecifications
| NF-H | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 200 kDa neurofilament protein, KIAA0845, NEFH, Neurofilament Heavy (200kDa), neurofilament heavy polypeptide, Neurofilament triplet H protein, neurofilament, heavy polypeptide, neurofilament, heavy polypeptide 200kDa, NF200, NFH, NF-H | |
| Human NF-H peptide, Accession # P12036 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1016103 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| P12036 | |
| 4744 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title