missing translation for 'onlineSavingsMsg'
Learn More

Human NFAM1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18717699
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18717699 25 μg 25µg
18130511 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18717699 Supplier R&D Systems Supplier No. MAB4046SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NFAM1 Monoclonal specifically detects NFAM1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NFAM1
Applications Western Blot
Classification Monoclonal
Clone 374301
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NET5
Gene Alias calcinerin/NFAT-activating ITAM-containing protein, Calcineurin/NFAT-activating ITAM-containing protein, CNAIP, CNAIPbK126B4.4, FLJ40652, NFAT activating protein with ITAM motif 1, NFAT activation molecule 1, NFAT-activating protein with ITAM motif 1
Gene Symbols NFAM1
Host Species Mouse
Immunogen E. coli-derived recombinant human NFAM1 Gln43-Lys163 Accession # Q8NET5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 150372
Test Specificity Detects human NFAM1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.