Learn More
Human NFIL3/E4BP4 Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB11657-SP
 View more versions of this product
Product Code. 30484814
  • 232.00 € / 25µg
Description
NFIL3 Monoclonal antibody specifically detects NFIL3 in Human samples. It is validated for Western Blot,Simple Western
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NFIL3
Monoclonal
Unconjugated
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2μm filtered solution in PBS.
E4BP4, E4BP4IL3BP1, IL3BP1, Interleukin-3 promoter transcriptional activator, Interleukin-3-binding protein 1, NF-IL3A, NFIL3AE4 promoter-binding protein 4, nuclear factor interleukin 3 regulated protein, nuclear factor interleukin-3-regulated protein, nuclear factor, interleukin 3 regulated, Transcriptional activator NF-IL3A
E. coli- derived recombinant human NFIL-3, Lys140-Gly462, Accession # Q16649
25 μg
Primary
Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Purified
Western Blot, Simple Western
1095812
Western Blot 2 μg/mL, Simple Western 20 μg/mL
Q16649
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4783
Human
Antibody
IgG2a