missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NGL-3/LRRC4B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB69191-SP
This item is not returnable.
View return policy
Description
NGL-3/LRRC4B/LRCH4B Monoclonal specifically detects NGL-3/LRRC4B/LRCH4B in Human samples. It is validated for Western Blot.
Specifications
| NGL-3/LRRC4B/LRCH4B | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NT99 | |
| LRRC4B | |
| Chinese hamster ovary cell line CHO-derived recombinant human NGL-3/LRRC4B Ala47-Lys576 Accession # Q9NT99 | |
| 25 μg | |
| Primary | |
| Detects human NGL-3/LRRC4B in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) LRRC3, rhLRRC4, or rhLRRC32 is observed. | |
| Human | |
| Purified |
| Western Blot | |
| 702008 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp761A179, HSM, Leucine Rich Repeat Containing 4B, Leucine-Rich Repeat-Containing Protein 4B, Leucine-Rich Repeats And Immunoglobulin-Like Domains 4, LRIG4, LRRC4B, Netrin-G3 Ligand, NGL3, NGL-3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 94030 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction