missing translation for 'onlineSavingsMsg'
Learn More

Human Nidogen-1/Entactin Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

208.00 € - 2040.00 €

Specifications

Antigen Nidogen-1/Entactin
Clone 302117
Dilution Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18712847
GREENER_CHOICE
View Documents
R&D Systems
MAB2570-SP
25 μg
208.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18749658
GREENER_CHOICE
View Documents
R&D Systems
MAB2570-100
100 μg
517.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18714137
GREENER_CHOICE
View Documents
R&D Systems
MAB2570-500
500 μg
2040.00 €
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nidogen-1/Entactin Monoclonal specifically detects Nidogen-1/Entactin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA Capture (Matched Antibody Pair).
Specifications

Specifications

Nidogen-1/Entactin
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
enactin, Entactin, Entactin-1, NID1, NID-1, NIDentactin, nidogen (enactin), nidogen 1, nidogen-1, Nidogen1
NID1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
302117
Western Blot, Immunohistochemistry, Immunocytochemistry, ELISA
Unconjugated
Mouse
Human
AAH45606.1
4811
Mouse myeloma cell line NS0-derived recombinant human Nidogen-1/Entactin Leu29-Lys1114 (Gln1113Arg) Accession # AAH45606.1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Nidogen-1/Entactin in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Nidogen-2 is observed.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.