missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKG2D/CD314 Alexa Fluor™ 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
213.00 € - 507.00 €
Specifications
| Antigen | NKG2D/CD314 |
|---|---|
| Clone | 149810 |
| Dilution | Flow Cytometry 5 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722149
![]() |
R&D Systems
FAB139N-025 |
25 Tests |
213.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18727097
![]() |
R&D Systems
FAB139N-100 |
100 Tests |
507.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NKG2D/CD314 Monoclonal specifically detects NKG2D/CD314 in Human samples. It is validated for Flow Cytometry.Specifications
| NKG2D/CD314 | |
| Flow Cytometry 5 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 22914 | |
| BaF3 mouse pro-B cell line transfected with human NKG2D/CD314 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 149810 | |
| Flow Cytometry | |
| Alexa Fluor 700 | |
| Mouse | |
| Human | |
| CD314, CD314 antigen, D12S2489E, FLJ17759, FLJ75772, Killer cell lectin-like receptor subfamily K member 1, killer cell lectin-like receptor subfamily K, member 1, KLR, KLRK1, NK cell receptor D, NKG2-D, NKG2-D type II integral membrane protein, NKG2-D-activating NK receptor, NKG2DDNA segment on chromosome 12 (unique) 2489 expressed sequence | |
| KLRK1 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human NKG2D/CD314 in Western blots. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title