missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NKp44/NCR2 PE-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB22491P-025
This item is not returnable.
View return policy
Description
NKp44/NCR2 Monoclonal specifically detects NKp44/NCR2 in Human samples. It is validated for Flow Cytometry.
Specifications
| NKp44/NCR2 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD336, CD336 antigen, LY95, LY95lymphocyte antigen 95 homolog (activating NK-receptor; NK-p44), lymphocyte antigen 95 (activating NK-receptor; NK-p44), Lymphocyte antigen 95 homolog, natural cytotoxicity triggering receptor 2, Natural killer cell p44-related protein, NCR2, NK cell activating receptor (NKp44), NK cell-activating receptor, NKP44, NK-p44dJ149M18.1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9436 | |
| Human | |
| Purified |
| Flow Cytometry | |
| 253415 | |
| Flow Cytometry 10 uL/10^6 cells | |
| CAB52289 | |
| NCR2 | |
| Mouse myeloma cell line NS0-derived recombinant human NKp44/NCR2 Gln22-Pro190 Accession # CAB52289 | |
| 25 Tests | |
| Primary | |
| Detects human NKp44/NCR2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) NKp30, rhNKp46, rhNKp80, or recombinant mouse NKp46 is observed. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction