missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Notch-1 Intracellular Domain Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF3647-SP
This item is not returnable.
View return policy
Description
Notch-1 Polyclonal specifically detects Notch-1 in Human samples. It is validated for Western Blot, Flow Cytometry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry, CyTOF-ready.
Specifications
| Notch-1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.1.2.11, EC 3.4.21.68, hN1, Notch (Drosophila) homolog 1 (translocation-associated), notch 1Notch homolog 1, translocation-associated (Drosophila), Notch homolog 1, translocation-associated, Notch1, TAN1, TAN1neurogenic locus notch homolog protein 1, Translocation-associated notch protein TAN-1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4851 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| ChIP Assay, CyTOF, Flow Cytometry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| P46531 | |
| NOTCH1 | |
| E. coli-derived recombinant human Notch-1 Gly2428-Lys2556 Accession # P46531 | |
| 25 μg | |
| Primary | |
| Detects the intracellular domain (ICD) of human Notch-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Notch-2 ICD, rhNotch-3 ICD, and rhNotch-4 ICD is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction