missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Nox4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8158-100
This item is not returnable.
View return policy
Description
Nox4 Monoclonal specifically detects Nox4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| Nox4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| EC 1.6.3, Kidney oxidase-1, Kidney superoxide-producing NADPH oxidase, KOX, KOX-1, KOX-1Renal NAD(P)H-oxidase, NADPH oxidase 4, RENOX, RENOXEC 1.6.3.- | |
| E. coli-derived recombinant human NOX4, Lys450-Ser578, Accession No. Q9NPH5 | |
| 100 μg | |
| 50507.0 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG2a |
| Immunohistochemistry, Immunocytochemistry | |
| 996715 | |
| Immunohistochemistry 1-25 μg/mL, Immunocytochemistry 5-25 μg/mL | |
| Q9NPH5 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human NOX4 in direct ELISAs. | |
| Human | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction