Learn More
Human NTB-A/SLAMF6 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems FAB19081A-100
 View more versions of this product
Product Code. 18749336
  • 558.00 € / 100 tests
Description
NTB-A/SLAMF6/CD352 Monoclonal specifically detects NTB-A/SLAMF6/CD352 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NTB-A/SLAMF6/CD352
Monoclonal
APC
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Activating NK receptor, CD352, CD352 antigen, KALIFLJ50657, Ly108, NK-T-B-antigen, NTBA, NTBA receptor, NTB-AMGC104953, NTBAT- and B-cell antigen, SF2000, SLAM family member 6, SLAMF6
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
114836
Human
Purified
Flow Cytometry
292811
Flow Cytometry 10 uL/10^6 cells
Q96DU3
SLAMF6
Mouse myeloma cell line NS0-derived recombinant human NTB-A/SLAMF6 Leu28-Lys225 Accession # Q96DU3
100 Tests
Primary
Detects human NTB-A/SLAMF6 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BLAME or rhCRACC is observed.
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
IgG2a