missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ORM2 Antibody (Clone 959445), R&D Systems™

Mouse Monoclonal Antibody
188.00 € - 388.00 €
Specifications
| Antigen | Orosomucoid 2 |
|---|---|
| Clone | 959445 |
| Dilution | Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18753869
![]() |
R&D Systems
MAB9306-SP |
25 μg |
188.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18754668
![]() |
R&D Systems
MAB9306-100 |
100 μg |
388.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Orosomucoid 2 Monoclonal specifically detects Orosomucoid 2 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| Orosomucoid 2 | |
| Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 5-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| P19652 | |
| 5005 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 959445 | |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| AGP 2, AGP2, AGP2alpha-1-acid glycoprotein 2, AGP-B, AGP-B', alpha-1-acid glycoprotein, type 2, OMD 2, orosomucoid 2, orosomucoid-2 | |
| Chinese hamster ovary cell line CHO-derived recombinant human ORM-2 Met1-Glu201 Accession # P19652 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ORM-2 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title