missing translation for 'onlineSavingsMsg'
Learn More

Human Osteoprotegerin/TNFRSF11B Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18700387
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700387 500 μg 500µg
18778009 25 μg 25µg
18796468 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18700387 Supplier R&D Systems Supplier No. MAB805500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 4 publications

Osteoprotegerin/TNFRSF11B Monoclonal specifically detects Osteoprotegerin/TNFRSF11B in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Osteoprotegerin/TNFRSF11B
Applications Neutralization
Classification Monoclonal
Clone 69146
Conjugate Unconjugated
Dilution Neutralization 0.15-0.3 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAB53709
Gene Alias OCIF, OCIFMGC29565, OPG, OPGtumor necrosis factor receptor superfamily member 11B, Osteoclastogenesis inhibitory factor, Osteoprotegerin, TNFRSF11B, TR1, tumor necrosis factor receptor superfamily, member 11b
Gene Symbols TNFRSF11B
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Osteoprotegerin/TNFRSF11B Glu22-Leu401 Accession # AAB53709
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4982
Test Specificity Detects human Osteoprotegerin/TNFRSF11B in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Osteoprotegerin is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.