missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40 Ligand/TNFSF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
193.00 € - 429.00 €
Specifications
| Antigen | OX40 Ligand/TNFSF4 |
|---|---|
| Clone | 159410 |
| Dilution | ELISA |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18721217
![]() |
R&D Systems
MAB10544-SP |
25 μg |
193.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18792228
![]() |
R&D Systems
MAB10544-100 |
100 μg |
429.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OX40 Ligand/TNFSF4 Monoclonal specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for ELISA.Specifications
| OX40 Ligand/TNFSF4 | |
| ELISA | |
| Monoclonal | |
| Supernatant | |
| Human | |
| P23510 | |
| 7292.0 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
| 159410 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Mouse myeloma cell line NS0-derived recombinant human OX40 Ligand/TNFRSF4, Gln51-Leu183, Accession No. P23510 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human OX40 Ligand/TNFSF4 in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title