missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40 Ligand/TNFSF4 Antibody, R&D Systems™
Mouse Monoclonal Antibody
201.00 € - 453.00 €
Specifications
| Antigen | OX40 Ligand/TNFSF4 |
|---|---|
| Clone | 1034083 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18142621
|
R&D Systems
MAB10656-100 |
100 μg |
453.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18141561
|
R&D Systems
MAB10656-SP |
25 μg |
201.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OX40 Ligand/TNFSF4 Monoclonal antibody specifically detects OX40 Ligand/TNFSF4 in Human samples. It is validated for ImmunohistochemistrySpecifications
| OX40 Ligand/TNFSF4 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| CD134 ligand, CD134L, CD252, CD252 antigen, Glycoprotein Gp34, gp34, OX40 antigen ligand, OX40 ligand, OX40L, OX-40L, OX4OL, TAX transcriptionally-activated glycoprotein 1, tax-transcriptionally activated glycoprotein 1 (34kD), tax-transcriptionally activated glycoprotein 1, 34kD, TNFSF4, tumor necrosis factor (ligand) superfamily, member 4, tumor necrosis factor ligand superfamily member 4, TXGP1 | |
| Mouse myeloma cell line NS0-derived human OX40 Ligand/TNFRSF4, Gln51-Leu183, Accession # Q6FGS4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1034083 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q6FGS4 | |
| 7292 | |
| IgG2b | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 6 months, -20°C to -70°C under sterile conditions after reconstitution. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts