missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human OX40/TNFRSF4 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB10545-SP
This item is not returnable.
View return policy
Description
OX40/TNFRSF4 Monoclonal specifically detects OX40/TNFRSF4 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| OX40/TNFRSF4 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ACT-135, ACT35 antigen, ACT35ATC35 antigen, CD134, CD134 antigen, Ly-70, OX40 cell surface antigen, OX40 homologue, OX40L receptor, OX40lymphoid activation antigene ACT35, TAX transcriptionally-activated glycoprotein 1 receptor, tax-transcriptionally activated glycoprotein 1 receptor, TNFRSF4, tumor necrosis factor receptor superfamily member 4, tumor necrosis factor receptor superfamily, member 4, Txgp1, TXGP1L, TXGP1LOX40 antigen | |
| Mouse myeloma cell line, NS0-derived human OX40/TNFRSF4, Leu29-Ala216, Accession # P43489 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| Flow Cytometry, CyTOF | |
| 2553B | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P43489 | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 7293 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction