missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human P-Rex1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8076-SP
This item is not returnable.
View return policy
Description
P-Rex1 Monoclonal specifically detects P-Rex1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| P-Rex1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL | |
| Q8TCU6 | |
| PREX1 | |
| E. coli-derived recombinant human P-Rex1 Lys1313-Arg1434 Accession # Q8TCU6 | |
| 25 μg | |
| Primary | |
| Detects human P-Rex1 in ELISAs and Western Blots. | |
| Human | |
| Purified |
| Western Blot | |
| 869704 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1415, phosphatidylinositol 34,5-trisphosphate-dependent Rac exchanger 1 protein, phosphatidylinositol-34,5-trisphosphate-dependent Rac exchange factor 1, PRex1, ptdIns(34,5)-dependent Rac exchanger 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 57580 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction